6-Chloro-3H-spiro[isobenzofuran-1,4'-piperidine] hydrochloride 97%

Supplier: Ambeed
Warning

A169848-100MG A169848-250MG A169848-1G
103806-422EA 327 USD
103806-422 76985-370 76985-372 77081-782
6-Chloro-3H-spiro[isobenzofuran-1,4'-piperidine] hydrochloride 97%
6-Chloro-3H-spiro[isobenzofuran-1,4'-piperidine] hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₅Cl₂NO
MW: 260.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20921934
CAS Number: 1359703-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR