6-Chloro-3-fluoro-2-(trifluoromethyl)pyridine 98%

Supplier: Ambeed
Danger

A116114-100MG A116114-250MG A116114-1G
103798-676EA 242 USD
103798-676 103824-540 77009-184 77009-186 77297-422
6-Chloro-3-fluoro-2-(trifluoromethyl)pyridine 98%
6-Chloro-3-fluoro-2-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₂ClF₄N
Boiling Pt: 144.6±35.0 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD16610411
CAS Number: 1227511-58-1
UN: 1993
ADR: 3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR