6-Chloro-3,4-pyridinediamine 98%

Supplier: Ambeed
Warning

A350381-5G A350381-10G A350381-250MG
103826-036EA 39 USD
103826-036 103837-030 76840-136 76876-286 76876-288
6-Chloro-3,4-pyridinediamine 98%
6-Chloro-3,4-pyridinediamine

Precaución: for Research Use Only

Formula: C₅H₆ClN₃
MW: 143.5700 g/mol
Flash Pt: 201.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04037269
CAS Number: 89182-17-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR