6-Chloro-1-tetralone 97%

Supplier: Ambeed
Warning

Synonyms: 6-Chloro-3,4-dihydronaphthalen-1(2H)-one, 6-Chloro-3,4-dihydro-1(2H)-naphthalenone

A122173-250MG A122173-1G A122173-25G
103800-186EA 13 USD
103800-186 77161-348 77297-856 77566-302
6-Chloro-1-tetralone 97%
6-Chloro-1-tetralone

Precaución: For research use only.

Formula: C₁₀H₉ClO
MW: 180.63 g/mol
Boiling Pt: 308.6 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD04114377
CAS Number: 26673-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR