6-Chloro-3-(trifluoromethyl)pyridin-2-amine 95%

Supplier: Ambeed
Warning

A316436-1G A316436-5G A316436-100MG A316436-250MG
103825-102EA 74 USD
103825-102 103836-028 76838-870 76838-872 76977-984 76977-986
6-Chloro-3-(trifluoromethyl)pyridin-2-amine 95%
6-Chloro-3-(trifluoromethyl)pyridin-2-amine

Precaución: For research use only

Formula: C₆H₄ClF₃N₂
MW: 196.558 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD19707620
CAS Number: 79456-27-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR