6-Chloro-2-naphthoic acid 95%

Supplier: Ambeed
Warning

A150112-100MG A150112-250MG
103804-128EA 248 USD
103804-128 77010-712 77010-714
6-Chloro-2-naphthoic acid 95%
6-Chloro-2-naphthoic acid

Precaución: for Research Use Only

Formula: C₁₁H₇ClO₂
MW: 206.63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11848443
CAS Number: 5042-97-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR