6-Chloro-2-methylimidazo[1,2-b]pyridazine 95%

Supplier: Ambeed
Warning

A190045-10G A190045-250MG A190045-1G A190045-5G
103816-256EA 32 USD
103816-256 103846-852 76799-958 76799-960 77302-200 77302-202
6-Chloro-2-methylimidazo[1,2-b]pyridazine 95%
6-Chloro-2-methylimidazo[1,2-b]pyridazine

Precaución: for Research Use Only

Formula: C₇H₆ClN₃
MW: 167.596 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07778346
CAS Number: 14793-00-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR