2-Chloro-6-methyl-5-nitropyridine 98%

Supplier: Ambeed
Warning

Synonyms: 6-Chloro-2-methyl-3-nitropyridine, 2-Chloro-5-nitro-6-picoline, 6-Chloro-3-nitro-2-picoline

A161894-100G A161894-10G A161894-25G A161894-5G
103843-790EA 13 USD
103843-790 77286-478 77286-480 77319-802 77319-804
2-Chloro-6-methyl-5-nitropyridine 98%
2-Chloro-6-methyl-5-nitropyridine

Precaución: for Research Use Only

Formula: C₆H₅ClN₂O₂
Boiling Pt: 255.9 °C (760 mm Hg)
Melting Pt: 54…58 °C
Flash Pt: 108.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00234160
CAS Number: 22280-60-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR