6-Chloro-2-methoxynicotinonitrile 97%

Supplier: Ambeed
Warning

A202586-100MG A202586-250MG
103817-404EA 135 USD
103817-404 103848-048 76986-506 76986-508
6-Chloro-2-methoxynicotinonitrile 97%
6-Chloro-2-methoxynicotinonitrile

Precaución: for Research Use Only

Formula: C₇H₅ClN₂O
MW: 168.58 g/mol
Boiling Pt: 272.1±35.0 °C (760 mm Hg)
Flash Pt: 118.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD13659332
CAS Number: 121643-46-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR