6-Chloro-2-methoxynicotinaldehyde 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Chloro-2-methoxypyridine-3-carbaldehyde
A141468-100MG
A141468-10G
A141468-250MG
A141468-1G
A141468-25G
103803-000EA
23
USD
103803-000
77162-326
77162-328
77162-330
77316-160
77316-162
6-Chloro-2-methoxynicotinaldehyde 95%
6-Chloro-2-methoxynicotinaldehyde
Precaución: for Research Use Only
Formula:
C₇H₆ClNO₂ MW: 171.581 g/mol Boiling Pt: 275.2±35.0 °C (760 mm Hg) Melting Pt: 62…64 °C Flash Pt: 120.2 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD11847276 CAS Number: 95652-81-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...