6-Chloro-2-methoxy-3-nitropyridine 95%
Supplier: Ambeed
|
Warning
|
A150343-100G
A150343-10G
A150343-250MG
A150343-1G
A150343-25G
A150343-5G
103804-146EA
23
USD
103804-146
103842-282
76768-132
76768-134
76768-136
77317-792
77317-794
77317-796
6-Chloro-2-methoxy-3-nitropyridine 95%
6-Chloro-2-methoxy-3-nitropyridine
Precaución: for Research Use Only
Formula:
C₆H₅ClN₂O₃ MW: 188.5700 g/mol Boiling Pt: 284.8 °C (760 mm Hg) Flash Pt: 126 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09033798 CAS Number: 40851-91-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...