6-Chloro-2-iodo-3-hydroxypyridine 98%

Supplier: Ambeed
Warning

Synonyms: 6-Chloro-2-iodopyridin-3-ol

A142874-250MG A142874-1G A142874-5G
103803-220EA 16 USD
103803-220 76767-886 77316-468 77316-470
6-Chloro-2-iodo-3-hydroxypyridine 98%
6-Chloro-2-iodo-3-hydroxypyridine

Precaución: for Research Use Only

Formula: C₅H₃ClIO
Boiling Pt: 332.821 °C (760 mm Hg)
Melting Pt: 191…193 °C
Flash Pt: 155.085 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07636749
CAS Number: 188057-26-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR