6-Chloro-2-hydroxynicotinic acid 95%

Supplier: Ambeed
Warning

A221272-100MG A221272-250MG A221272-5G
103818-910EA 46 USD
103818-910 103849-792 76814-904 76814-906 77088-392
6-Chloro-2-hydroxynicotinic acid 95%
6-Chloro-2-hydroxynicotinic acid

Precaución: for Research Use Only

Formula: C₆H₄ClNO₃
MW: 173.55 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18343615
CAS Number: 38076-76-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR