6-Chloro-2-fluoropyridin-3-ol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Chloro-2-fluoro-3-hydroxypyridine
A237740-100MG
A237740-250MG
A237740-1G
103811-174EA
47
USD
103811-174
103850-956
76987-536
76987-538
77304-090
6-Chloro-2-fluoropyridin-3-ol 98%
6-Chloro-2-fluoropyridin-3-ol
Precaución: for Research Use Only
Formula:
C₅H₃ClFNO MW: 147.535 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07437887 CAS Number: 883107-68-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...