6-Chloro-2,5-dimethylpyrimidin-4-amine 95%

Supplier: Ambeed
Warning

A260911-250MG A260911-1G
103812-510EA 17 USD
103812-510 76988-188 77091-992
6-Chloro-2,5-dimethylpyrimidin-4-amine 95%
6-Chloro-2,5-dimethylpyrimidin-4-amine

Precaución: for Research Use Only

Formula: C₆H₈ClN₃
MW: 157.6 g/mol
Boiling Pt: 279.7 °C (760 mm Hg)
Flash Pt: 123 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01689894
CAS Number: 18260-92-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR