6-Chloro-2,5-dimethylpyridazin-3(2H)-one 98%

Supplier: Ambeed
Warning

A116203-100MG A116203-250MG
103798-692EA 60 USD
103798-692 77009-188 77009-190
6-Chloro-2,5-dimethylpyridazin-3(2H)-one 98%
6-Chloro-2,5-dimethylpyridazin-3(2H)-one

Precaución: for Research Use Only

Formula: C₆H₇ClN₂O
MW: 158.59 g/mol
Boiling Pt: 222.7±23.0 °C (760 mm Hg)
Flash Pt: 88.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD27923628
CAS Number: 1114563-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR