6-Chloro-2,3-dimethylpyridine 95%

Supplier: Ambeed
Warning

A102825-100MG A102825-250MG
103796-428EA 113 USD
103796-428 103822-126 76982-236 76982-238
6-Chloro-2,3-dimethylpyridine 95%
6-Chloro-2,3-dimethylpyridine

Precaución: for Research Use Only

Formula: C₇H₈ClN
MW: 141.598 g/mol
Boiling Pt: 210.1 °C (760 mm Hg)
Flash Pt: 100.6 °C
Storage Temperature: Ambient
MDL Number: MFCD11505294
CAS Number: 72093-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR