6-Chloro-2,3-dihydro-1H-pyrrolo[3,2-c]pyridine 95%

Supplier: Ambeed
Warning

A320809-100MG A320809-250MG
103825-250EA 398 USD
103825-250 103836-182 76978-070 76978-072
6-Chloro-2,3-dihydro-1H-pyrrolo[3,2-c]pyridine 95%
6-Chloro-2,3-dihydro-1H-pyrrolo[3,2-c]pyridine

Precaución: for Research Use Only

Formula: C₇H₇ClN₂
MW: 154.6 g/mol
Boiling Pt: 287.2 °C (760 mm Hg)
Flash Pt: 127.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00980787
CAS Number: 23596-25-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR