6-Chloro-2,3-dihydro-1H-indole 95%

Supplier: Ambeed
Warning

A105073-100MG A105073-250MG A105073-1G A105073-5G
103796-832EA 112 USD
103796-832 103822-558 77160-436 77160-438 77309-428 77309-430
6-Chloro-2,3-dihydro-1H-indole 95%
6-Chloro-2,3-dihydro-1H-indole

Precaución: for Research Use Only

Formula: C₈H₈ClN
MW: 153.6 g/mol
Boiling Pt: 250.3 °C (760 mm Hg)
Flash Pt: 105.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07371639
CAS Number: 52537-00-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR