6-Chloro-2-(trifluoromethyl)nicotinonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 6-Chloro-2-(trifluoromethyl)-3-pyridinecarbonitrile

A152090-100MG A152090-250MG A152090-1G
103804-348EA 130 USD
103804-348 103842-504 77162-852 77162-854 77318-114
6-Chloro-2-(trifluoromethyl)nicotinonitrile 98%
6-Chloro-2-(trifluoromethyl)nicotinonitrile

Precaución: Inert atmosphere, 2…8 °C.

Formula: C₇H₂ClF₃N₂
MW: 206.552 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11848371
CAS Number: 1245913-20-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR