6-Chloro-2-(trifluoromethyl)nicotinic acid 95%

Supplier: Ambeed
Warning

A163084-100MG A163084-250MG A163084-1G A163084-5G
103805-682EA 136 USD
103805-682 103843-940 76824-132 76824-134 77300-630 77300-632
6-Chloro-2-(trifluoromethyl)nicotinic acid 95%
6-Chloro-2-(trifluoromethyl)nicotinic acid

Precaución: for Research Use Only

Formula: C₇H₃ClF₃NO₂
Boiling Pt: 281.7 °C (760 mm Hg)
Flash Pt: 124.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11840271
CAS Number: 261635-83-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR