6-Chloro-1H-pyrrolo[3,2-c]pyridine 97%

Supplier: Ambeed
Warning

Synonyms: 6-Chloro-5-azaindole

A241081-100MG A241081-10G A241081-250MG A241081-1G A241081-25G A241081-5G
103811-390EA 19 USD
103811-390 103851-196 76771-420 76771-422 76771-424 77090-352 77090-354 77090-356
6-Chloro-1H-pyrrolo[3,2-c]pyridine 97%
6-Chloro-1H-pyrrolo[3,2-c]pyridine

Precaución: for Research Use Only

Formula: C₇H₅ClN₂
MW: 152.5800 g/mol
Boiling Pt: 335.8 °C (760 mm Hg)
Flash Pt: 187.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08690133
CAS Number: 74976-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR