6-Chloro-1H-pyrrolo[2,3-b]pyridine 95%
Supplier: Ambeed
|
Danger
|
Synonyms:
6-Chloro-7-azaindole
A144512-100G
A144512-10G
A144512-250MG
A144512-25G
A144512-5G
A144512-1G
103803-414EA
7
USD
103803-414
103841-490
76767-952
76767-954
76767-956
77299-392
77299-394
77316-738
6-Chloro-1H-pyrrolo[2,3-b]pyridine 95%
6-Chloro-1H-pyrrolo[2,3-b]pyridine
Precaución: for Research Use Only
Formula:
C₇H₅ClN₂ MW: 152.58 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD06659666 CAS Number: 55052-27-2 UN: 1759 ADR: 8,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...