6-Chloro-1H-pyrazolo[4,3-c]pyridine 98%

Supplier: Ambeed
Warning

A245028-100G A245028-10G A245028-250MG A245028-1G A245028-25G A245028-5G
103811-620EA 14 USD
103811-620 103851-454 76976-564 76976-566 76976-568 77090-684 77090-686 77090-688
6-Chloro-1H-pyrazolo[4,3-c]pyridine 98%
6-Chloro-1H-pyrazolo[4,3-c]pyridine

Precaución: for Research Use Only

Formula: C₆H₄ClN₃
MW: 153.5700 g/mol
Boiling Pt: 358 °C (760 mm Hg)
Melting Pt: 220…224 °C
Flash Pt: 201 °C
Storage Temperature: Ambient
MDL Number: MFCD17010105
CAS Number: 1206979-33-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR