6-Chloro-1H-pyrazolo[3,4-d]pyrimidine 97%

Supplier: Ambeed
Warning

A189379-100MG A189379-250MG A189379-1G
103816-182EA 30 USD
103816-182 103846-774 76784-862 76784-864 77084-546
6-Chloro-1H-pyrazolo[3,4-d]pyrimidine 97%
6-Chloro-1H-pyrazolo[3,4-d]pyrimidine

Precaución: for Research Use Only

Formula: C₅H₃ClN₄
MW: 154.5571 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD10697157
CAS Number: 23002-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR