6-Chloro-1H-indazole-4-carboxylic acid 95%

Supplier: Ambeed
Warning

A476572-1G A476572-100MG A476572-250MG
103828-084EA 205 USD
103828-084 76843-080 76990-950 76990-952
6-Chloro-1H-indazole-4-carboxylic acid 95%
6-Chloro-1H-indazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₅ClN₂O₂
MW: 196.59 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07781757
CAS Number: 885522-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR