6-Chloro-1H-indazole-3-carboxaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: 6-Chloro-1H-indazole-3-carbaldehyde

A353603-100MG A353603-250MG A353603-1G A353603-5G
103826-100EA 39 USD
103826-100 103837-102 76828-634 76828-636 76840-216 76840-218
6-Chloro-1H-indazole-3-carboxaldehyde 95%
6-Chloro-1H-indazole-3-carboxaldehyde

Precaución: for Research Use Only

Formula: C₈H₅ClN₂O
MW: 180.591 g/mol
Boiling Pt: 391.7±22.0 °C (760 mm Hg)
Flash Pt: 190.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07781580
CAS Number: 885521-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR