6-Chloro-1H-indazole 98%

Supplier: Ambeed
Warning

A158114-100MG A158114-10G A158114-250MG A158114-1G A158114-5G A158114-25G
103805-096EA 9 USD
103805-096 103843-304 77163-078 77163-080 77163-082 77319-146 77319-148 77683-320
6-Chloro-1H-indazole 98%
6-Chloro-1H-indazole

Precaución: For research use only.

Formula: C₇H₅ClN₂
MW: 152.5800 g/mol
Boiling Pt: 309.5 °C (760 mm Hg)
Flash Pt: 169.9 °C
Storage Temperature: Ambient
MDL Number: MFCD07774234
CAS Number: 698-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR