4-Amino-6-chloro-1H-indazole 97%

Supplier: Ambeed
Warning

Synonyms: 6-Chloro-1H-indazol-4-amine

A446199-1G A446199-100MG A446199-250MG
103827-738EA 139 USD
103827-738 76842-596 77684-224 77684-225
4-Amino-6-chloro-1H-indazole 97%
4-Amino-6-chloro-1H-indazole

Precaución: For research use only.

Formula: C₇H₆ClN₃
MW: 167.6 g/mol
Boiling Pt: 415.5±25.0 °C (760 mm Hg)
Flash Pt: 205.1 °C
Storage Temperature: Ambient
MDL Number: MFCD07781370
CAS Number: 885519-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR