6-Chloro-1,3-dimethyluracil 97%

Supplier: Ambeed
Warning

Synonyms: 6-Chloro-1,3-dimethylpyrimidine-2,4(1H,3H)-dione, 1,3-Dimethyl-6-chlorouracil

A300509-25G A300509-5G A300509-100G A300509-10G A300509-1G
103814-276EA 9 USD
103814-276 103835-426 76838-110 76838-112 77114-466 77114-468 77305-810
6-Chloro-1,3-dimethyluracil 97%
6-Chloro-1,3-dimethyluracil

Precaución: for Research Use Only

Formula: C₆H₇ClN₂O₂
MW: 174.59 g/mol
Boiling Pt: 224.6 °C (760 mm Hg)
Melting Pt: 109…113 °C
Flash Pt: 89.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00038066
CAS Number: 6972-27-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR