6-Bromothieno[2,3-d]pyrimidine 98%

Supplier: Ambeed
Warning

A112918-100MG A112918-250MG A112918-1G
103798-130EA 449 USD
103798-130 103823-964 76982-822 76982-824 77310-796
6-Bromothieno[2,3-d]pyrimidine 98%
6-Bromothieno[2,3-d]pyrimidine

Precaución: for Research Use Only

Formula: C₆H₃BrN₂S
MW: 215.07 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD22038097
CAS Number: 60703-80-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR