6-Bromo-2-hydroxyquinoxaline 95%

Supplier: Ambeed
Warning

A255780-100MG A255780-250MG A255780-1G
103812-212EA 57 USD
103812-212 103852-072 77031-682 77031-684 77091-532
6-Bromo-2-hydroxyquinoxaline 95%
6-Bromo-2-hydroxyquinoxaline

Precaución: for Research Use Only

Formula: C₈H₅BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD09834132
CAS Number: 55687-34-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR