6-Bromoquinoline-4-carboxylic acid 98%

Supplier: Ambeed
Warning

A134844-100G A134844-100MG A134844-10G A134844-250MG A134844-25G A134844-1G A134844-5G
103802-054EA 18 USD
103802-054 77027-256 77027-258 77027-260 77027-262 77298-736 77314-832 77314-834
6-Bromoquinoline-4-carboxylic acid 98%
6-Bromoquinoline-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₆BrNO₂
MW: 252.0600 g/mol
Flash Pt: 197.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00681026
CAS Number: 160233-76-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR