6-Bromoquinoline-3-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Bromo-3-carboxy-1-azanaphthalene, 6-Bromo-3-carboxyquinoline
A102967-10G
A102967-250MG
A102967-25G
A102967-1G
A102967-5G
103796-456EA
74
USD
103796-456
103822-160
76780-578
76780-580
76780-582
77309-026
77309-028
6-Bromoquinoline-3-carboxylic acid 97%
6-Bromoquinoline-3-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₀H₆BrNO₂ MW: 252.064 g/mol Boiling Pt: 403.1±30.0 °C (760 mm Hg) Melting Pt: 234…239 °C Flash Pt: 197.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD08437553 CAS Number: 798545-30-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...