6-Bromoquinoline 98%

Supplier: Ambeed
Warning

A274130-100G A274130-10G A274130-500G A274130-25G A274130-5G
103853-110EA 5 USD
103853-110 76692-900 76692-902 76692-904 77092-928 77092-930
6-Bromoquinoline 98%
6-Bromoquinoline

Precaución: for Research Use Only

Formula: C₉H₆BrN
MW: 208.06 g/mol
Boiling Pt: 278 °C (760 mm Hg)
Melting Pt: 24 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00024023
CAS Number: 5332-25-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR