6-Bromoquinolin-8-ol 95%

Supplier: Ambeed
Warning

A149696-100MG A149696-10G A149696-250MG A149696-5G
103804-072EA 83 USD
103804-072 103842-198 76823-794 76823-796 76823-798 77317-700
6-Bromoquinolin-8-ol 95%
6-Bromoquinolin-8-ol

Precaución: for Research Use Only

Formula: C₉H₆BrNO
MW: 224.05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11878495
CAS Number: 139399-64-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR