6-Bromoquinolin-4-ol 98%

Supplier: Ambeed
Warning

Synonyms: 6-Bromo-4-quinolinol, 6-Bromo-4-hydroxyquinoline

A356865-1G A356865-25G A356865-5G A356865-100G A356865-250MG
103826-178EA 7 USD
103826-178 103837-184 76840-326 76840-328 76840-330 77263-948 77263-950
6-Bromoquinolin-4-ol 98%
6-Bromoquinolin-4-ol

Precaución: for Research Use Only

Formula: C₉H₆BrNO
MW: 224.0540 g/mol
Boiling Pt: 321.7 °C (760 mm Hg)
Flash Pt: 148.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00272435
CAS Number: 145369-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR