6-Bromoquinolin-4(1H)-one 97%

Supplier: Ambeed
Warning

A223428-10G A223428-250MG A223428-1G A223428-25G A223428-5G
103819-036EA 21 USD
103819-036 103849-916 77030-882 77030-884 77088-536 77088-538 77088-540
6-Bromoquinolin-4(1H)-one 97%
6-Bromoquinolin-4(1H)-one

Precaución: for Research Use Only

Formula: C₉H₆BrNO
Boiling Pt: 321.7±42.0 °C (760 mm Hg)
Melting Pt: 282…284 °C
Flash Pt: 148.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07644510
CAS Number: 332366-57-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR