6-Bromoquinolin-2(1H)-one 96%
Supplier: Ambeed
|
Warning
|
A189513-1G
A189513-5G
A189513-25G
A189513-250MG
103816-196EA
14
USD
103816-196
103846-788
77084-560
77084-562
77302-180
77593-210
6-Bromoquinolin-2(1H)-one 96%
6-Bromoquinolin-2(1H)-one
Precaución: for Research Use Only
Formula:
C₉H₆BrNO MW: 224.05 g/mol Boiling Pt: 393.8±42.0 °C (760 mm Hg) Melting Pt: 278…280 °C Storage Temperature: Ambient |
MDL Number:
MFCD08703169 CAS Number: 1810-66-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...