6-Bromoquinazoline-2,4(1H,3H)-dione 98%

Supplier: Ambeed
Warning

A130655-100G A130655-100MG A130655-10G A130655-250MG A130655-1G A130655-25G A130655-5G
103801-448EA 13 USD
103801-448 76767-212 76767-214 76767-216 76767-218 77298-418 77313-900 77313-902
6-Bromoquinazoline-2,4(1H,3H)-dione 98%
6-Bromoquinazoline-2,4(1H,3H)-dione

Precaución: for Research Use Only

Formula: C₈H₅BrN₂O₂
MW: 241.04 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00462868
CAS Number: 88145-89-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR