6-Bromoquinazoline 96%

Supplier: Ambeed
Warning

A214643-25G A214643-5G A214643-100MG A214643-10G A214643-250MG A214643-1G
103818-422EA 17 USD
103818-422 103849-126 77087-656 77087-658 77152-560 77152-562 77152-564 77303-358
6-Bromoquinazoline 96%
6-Bromoquinazoline

Precaución: for Research Use Only

Formula: C₈H₅BrN₂
Boiling Pt: 304.2 °C (760 mm Hg)
Flash Pt: 137.808 °C
Storage Temperature: Ambient
MDL Number: MFCD08236695
CAS Number: 89892-21-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR