6-Bromoquinazolin-4-amine 98%
Supplier: Ambeed
|
Warning
|
A329100-250MG
A329100-1G
A329100-5G
103825-512EA
43
USD
103825-512
103836-466
76773-064
76839-434
76839-436
6-Bromoquinazolin-4-amine 98%
6-Bromoquinazolin-4-amine
Precaución: for Research Use Only
Formula:
C₈H₆BrN₃ MW: 224.0600 g/mol Boiling Pt: 385.4 °C (760 mm Hg) Storage Temperature: Refrigerator |
MDL Number:
MFCD02228849 CAS Number: 21419-48-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...