6-Bromopyrido[2,3-d]pyrimidin-4(1H)-one 98%

Supplier: Ambeed
Warning

A114693-10G A114693-250MG A114693-25G A114693-1G A114693-5G
103798-450EA 74 USD
103798-450 103824-294 76781-310 76781-312 76781-314 77311-108 77311-110
6-Bromopyrido[2,3-d]pyrimidin-4(1H)-one 98%
6-Bromopyrido[2,3-d]pyrimidin-4(1H)-one

Precaución: for Research Use Only

Formula: C₇H₄BrN₃O
MW: 226.03 g/mol
Boiling Pt: 396 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD11040199
CAS Number: 155690-79-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR