6-Bromopyridine-3,4-diamine 98%

Supplier: Ambeed
Warning

A399848-5G A399848-100MG A399848-250MG
103827-040EA 248 USD
103827-040 103838-074 76841-520 76990-228 76990-230
6-Bromopyridine-3,4-diamine 98%
6-Bromopyridine-3,4-diamine

Precaución: for Research Use Only

Formula: C₅H₆BrN₃
MW: 188.025 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11100655
CAS Number: 1033203-41-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR