2-Bromo-5-hydroxypyridine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Bromo-5-pyridinol, 6-Bromopyridin-3-ol
A135929-100G
A135929-10G
A135929-250MG
A135929-1G
A135929-25G
A135929-5G
103802-244EA
6
USD
103802-244
76972-346
76972-348
76972-350
77298-830
77298-832
77315-126
2-Bromo-5-hydroxypyridine 98%
2-Bromo-5-hydroxypyridine
Precaución: for Research Use Only
Formula:
C₅H₄BrNO Boiling Pt: 373 °C (760 mm Hg) Melting Pt: 135.5…136.5 °C Flash Pt: 179.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD04114184 CAS Number: 55717-45-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...