5-Amino-2-bromopyridine 97%

Supplier: Ambeed
Warning

Synonyms: 6-Bromopyridin-3-amine, 2-Bromopyridin-5-amine, 3-Amino-6-bromopyridine, 2-Bromo-5-pyridylamine

A147911-100G A147911-10G A147911-500G A147911-1G A147911-25G A147911-5G
103803-846EA 5 USD
103803-846 103841-960 77272-858 77272-860 77272-862 77299-612 77317-376 77317-378
5-Amino-2-bromopyridine 97%
5-Amino-2-bromopyridine

Precaución: for Research Use Only

Formula: C₅H₅BrN₂
MW: 173.01 g/mol
Boiling Pt: 291.2 °C (760 mm Hg)
Melting Pt: 75 °C
Storage Temperature: Ambient
MDL Number: MFCD01646116
CAS Number: 13534-97-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR