2-Amino-6-bromopyridine 98%

Supplier: Ambeed
Warning

Synonyms: 6-Bromo-2-pyridylamine, 6-Bromopyridin-2-amine

A131355-100G A131355-10G A131355-500G A131355-1G A131355-25G A131355-1000G A131355-5G
103808-424EA 437 USD
103808-424 103801-570 77009-968 77009-970 77009-972 77298-474 77298-476 77314-068 77314-070
2-Amino-6-bromopyridine 98%
2-Amino-6-bromopyridine

Precaución: for Research Use Only

Formula: C₅H₅BrN₂
MW: 173.01 g/mol
Boiling Pt: 272.9 °C (760 mm Hg)
Melting Pt: 88…91 °C
Flash Pt: 118.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00137843
CAS Number: 19798-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR