6-Bromo-3-pyridazinol 98%

Supplier: Ambeed
Warning

A236187-10G A236187-1G A236187-25G A236187-5G A236187-100G A236187-250MG
103811-076EA 7 USD
103811-076 103850-844 77013-660 77089-886 77089-888 77089-890 77890-369 77890-370
6-Bromo-3-pyridazinol 98%
6-Bromo-3-pyridazinol

Precaución: for Research Use Only

Formula: C₄H₃BrN₂O
Boiling Pt: 363.6 °C (760 mm Hg)
Melting Pt: 157…161 °C
Flash Pt: 173.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11223475
CAS Number: 51355-94-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR