6-Bromopyrazolo[1,5-a]pyrimidine 97%

Supplier: Ambeed
Warning

A104846-10G A104846-250MG A104846-25G A104846-1G A104846-5G
103796-786EA 7 USD
103796-786 103822-512 77025-366 77025-368 77025-370 77309-370 77309-372
6-Bromopyrazolo[1,5-a]pyrimidine 97%
6-Bromopyrazolo[1,5-a]pyrimidine

Precaución: for Research Use Only

Formula: C₆H₄BrN₃
MW: 198.02 g/mol
Melting Pt: 122…124 °C
Storage Temperature: Ambient
MDL Number: MFCD09832894
CAS Number: 705263-10-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR