6-Bromonicotinonitrile 96%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-5-cyanopyridine

A153773-100G A153773-1G A153773-10G
103804-562EA 6 USD
103804-562 103842-740 77273-064 77300-030 77748-057
6-Bromonicotinonitrile 96%
6-Bromonicotinonitrile

Precaución: for Research Use Only

Formula: C₆H₃BrN₂
MW: 183.01 g/mol
Boiling Pt: 261.6 °C (760 mm Hg)
Melting Pt: 136…140 °C
Flash Pt: 112 °C
Storage Temperature: Ambient
MDL Number: MFCD00234141
CAS Number: 139585-70-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR